ContactUsLinkComponent
Imhoff Cone and Cone Racks, Wheaton, DWK Life Sciences
Imhoff Cone and Cone Racks, Wheaton, DWK Life Sciences
Catalog # 80080-902
Small Business Enterprise
undefined
Imhoff Cone and Cone Racks, Wheaton, DWK Life Sciences
Catalog # 80080-902
Supplier Number:  W990800
Small Business Enterprise
Specifications
  • Length:
    45.1 cm (173/4")
  • Description:
    Imhoff Cone
  • Top Ø:
    10.8 cm (41/4")
  • Cat. no.:
    80080-902
  • Supplier No.:
    W990800
Specifications
About this item
Imhoff cone for volumetric determination of settleable solids in water and wastewater.
Styrene-acrylonitrile cone is externally graduated from plug up to 1000 ml: 0 - 1 ml in 0.1 ml; 1 - 10 ml in 0.5 ml; 10 - 40 ml in 1 ml; 40 - 100 ml in 2 ml; lines and numbers at 250, 500 and 1000 ml. Easily cleaned by removing the low-density polyethylene leak-free plug