KONTES® Soil Analysis Tubes, Kimble Chase
19×150 mm
Catalog # KT591175-0715
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 591175-0715
KONTES® Soil Analysis Tubes, Kimble Chase
Catalog # KT591175-0715
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
These test tubes are designed for use with Tekmar® 2016/2032 autosamplers and 4100/4200 automatic samplers equipped with 1.9 cm (³/₄") mounts.
Tubes feature a large opening to permit easy sample loading and to facilitate the weighing of solid and soil samples.
Documents
Kimble Petrochemical Glassware Brochure
Product Literature
Specifications
- Ø×L:19×150 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



