ContactUsLinkComponent
Synthware Tubes, Craig Recrystallization, Microscale, Kemtech America
Synthware Tubes, Craig Recrystallization, Microscale, Kemtech America
Catalog # 60004-148
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Tubes, Craig Recrystallization, Microscale, Kemtech America
Catalog # 60004-148
Supplier:  Kemtech America
Supplier Number:  T140002
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    2 mL, Complete
  • Environmentally Preferable:
  • Cat. no.:
    60004-148
  • Supplier No.:
    T140002
Specifications
About this item
Craig recystalllization tubes for microscale applications.
  • 3.3 Expansion borosilicate glass
  • Complete set
Complete tube includes a glass outer tube and an inner plug.