ContactUsLinkComponent
InLab® Redox Micro Electrode, METTLER TOLEDO®
  CA97001-694
undefined
InLab® Redox Micro Electrode, METTLER TOLEDO®
  CA97001-694
 :  51343203
  • Temperature Range:
    0...100 °C
  • Length:
    100 mm
  • Material:
    Glass
  • Cat. no.:
    CA97001-694
  • Internal Reference:
    ARGENTHAL™
  • Junction:
    Ceramic
  • Supplier No.:
    51343203
  • Electrolyte:
    3M KCl
Electrode measures oxidation/reduction power of a solution
  • Platinum ring indicator
  • Safelock™ closing ring
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head connector
Features a platinum ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters. Diameter: 6mm.