Order Entry
Canada
Orders LinkContactUsLinkComponent
InLab® Combination pH Electrode, METTLER TOLEDO®

InLab® Combination pH Electrode, METTLER TOLEDO®

Supplier:   Mettler Toledo
InLab® Combination pH Electrode, METTLER TOLEDO®
Supplier:  Mettler Toledo
Certificates
InLab® Combination pH Electrode, METTLER TOLEDO®
InLab® Combination pH Electrode, METTLER TOLEDO®

3 Options Available Below

Product Details & Documents

Electrode is a part of INGOLD tradition, there is no pH application for which there is not an optimal technical solution.

  • INGOLD tradition
  • S7 screw head connector

These electrodes cover the majority of applications, such as food processing, paper and checmical industries. For U402-611-DPA there is a specially developed thermostat controllable container for high precision measurements in automated cycles. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.

Specifications for Product Family

Specifications

  • Catalog No:
  • CA11238-656
  • 11238-654
  • CA97001-682
  • CA11238-648
  • Ø:
  • 7 mm
  • 12 mm
  • 3 mm

Specifications

Product Family Options

Product Information

  • Type
    For use with
    Length
    Fill solution
    Body material
    Temperature range
    pH range
    Internal reference
    Junction
    Availability
    Price
  • U402-611-DPA-S7/40
    40 mm
    DPA gel
    Glass
    0...80 °C (32...176 °F)
    1 - 11
    SteadyForce™
    Double, ceramic
    Log in for availability
    Log in for availability
    Each
    Each - $980.00
    $980.00
    Each
       $980.00

    Supplier Number: 59902917
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    11238-654
    Ø
    7 mm
    Product available on GSA Advantage®
  • 405-60-T-S7/120/9848
    120 mm
    FRISCOLYT-B
    Glass
    0...100 °C (32...212 °F)
    0 - 12
    ARGENTHAL
    Ceramic
    Log in for availability
    Log in for availability
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00

    Supplier Number: 59904591
    Specifications:
    More
    Specifications
    Cat. No.
    CA97001-682
    Ø
    12 mm
  • U402-M3-S7/200
    200 mm
    3 mol/L KCl
    Glass
    0...80 °C (32...176 °F)
    0 - 14
    Ag/AgCl
    Ceramic
    Log in for availability
    Log in for availability
    Each
    Each - $1,710.00
    $1,710.00
    Each
       $1,710.00

    Supplier Number: 59904572
    Specifications:
    More
    Specifications
    Cat. No.
    CA11238-648
    Ø
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.