To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Length:120 mm
- Temperature range:0 to 100°C
- Cat. no.:CA97001-670
- pH Range:0 - 14
- Body Material:Glass
- Supplier No.:51343195
Specifications
About this item
Electrode is recommended for applications in which the lifetime of the electrode is significantly shorter than that of a standard reference electrode
- High alkali glass body
- Dome-shaped membrane
- S7 screw head connector
High alkali glass body protects against corrosive and organic samples and the dome-shaped membrane is easy to clean. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.