ContactUsLinkComponent
InLab® Versatile Pro 3-in-1 pH Electrode, Replaceable Junction, METTLER TOLEDO®
InLab® Versatile Pro 3-in-1 pH Electrode, Replaceable Junction, METTLER TOLEDO®
Catalog # CA97001-656
Supplier:  Mettler Toledo
undefined
InLab® Versatile Pro 3-in-1 pH Electrode, Replaceable Junction, METTLER TOLEDO®
Catalog # CA97001-656
Supplier:  Mettler Toledo
Supplier Number:  51343031
Specifications
  • Temperature Range:
    0...100 °C
  • Length:
    120 mm
  • Cat. no.:
    CA97001-656
  • pH Range:
    0 - 14
  • Body Material:
    Polysulfone
  • Internal Reference:
    ARGENTHAL
  • Junction:
    Ceramic
  • Fill Solution:
    3M KCl
  • Supplier No.:
    51343031
Specifications
About this item
Electrode features a replaceable ceramic junction, which offers a solution to contaminated junctions without the higher cost or cleaning efforts required with open or sleeve junctions
  • Replaceable ceramic junction
  • Integrated temperature probe
  • Polysulfone shaft, universal glass membrane
  • ARGENTHAL™ lead-off and silver ion trap
  • BNC/RCA (Cinch) connector
Electrode also features integrated NTC 30 k[OMEGA] temperature probe, robust polysulfone shaft, universal glass membrane with low resistance, ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures, and BNC/RCA (Cinch) connector.