Order Entry
Canada
ContactUsLinkComponent
 
Synthware NMR Tube Washer, Kemtech America
Catalog # 60004-732
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware NMR Tube Washer, Kemtech America
Catalog # 60004-732
Supplier:  Kemtech America
Supplier Number:  T100024
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Synthware NMR Tube Washer
  • Cat. no.:
    60004-732
  • Supplier No.:
    T100024

Specifications

About this item

Tube washer features a [ST] 24/40 inner joint and 10 mm hose connection.

  • Designed for use with 5 mm NMR tubes only

The NMR tube is inserted into the tube cleaner with its plastic cap placed on the closed end to provide support and a vacuum seal. The washing solvents is sequentially added through the side tube while a vacuum is applied through the hose connection.