Order Entry
Canada
ContactUsLinkComponent
ORION® ROSS® Sure-Flow® Semi-Micro Combination pH Electrode, Thermo Scientific
ORION® ROSS® Sure-Flow® Semi-Micro Combination pH Electrode, Thermo Scientific
Catalog # CA10010-774
undefined
ORION® ROSS® Sure-Flow® Semi-Micro Combination pH Electrode, Thermo Scientific
Catalog # CA10010-774
Supplier Number:  8175BNWP

Specifications

  • Temperature Range:
    0...100 °C
  • Length:
    165 mm
  • Cat. no.:
    CA10010-774
  • pH Range:
    0 - 14
  • Body Material:
    Epoxy
  • Internal Reference:
    ROSS
  • Junction:
    Sure-flow
  • Connector:
    BNC
  • Supplier No.:
    8175BNWP

Specifications

About this item

Semi-micro electrode is ideal for pH measurements in test tubes and other small vessels.

Stable to 0.01 pH in 30 s. Features the unique Sure-Flow® liquid-to-liquid junction. Diameter: 6 mm. Cap diameter: 16 mm. Lead length: 1 m.