Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Gram's iodine solution

Supplier:   G-Biosciences
Gram's iodine solution
Supplier:  G-Biosciences
Certificates
Gram's iodine solution

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Gram Iodine Solution acts as a mordant .

  • An aqueous solution of Iodine

It is used in Gram staining to differentiate gram-positive from gram-negative bacteria

Synonyms:  Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
  • CAS Number:12298-68-9
  • Formula:I₃K
  • MDL Number:MFCD00134128
  • Molecular Weight:419.812 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    250 ml
    Log in for availability
    Log in for availability
    Each
    Each - $41.32
    $41.32
    Each
       $41.32

    Supplier Number: 786-2092
  • Plastic bottle
    5×250 ml
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $161.24
    $161.24
    Kit of 1
       $161.24

    Supplier Number: 786-2093
  • Plastic bottle
    1 Gallon
    Log in for availability
    Log in for availability
    Each
    Each - $319.48
    $319.48
    Each
       $319.48

    Supplier Number: 786-2094

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.