PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
Supplier: Corning
PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
Supplier: Corning
Certificates
About this item
PYREXPLUS®, borosilicate glass.
- Fluoropolymer resin stopcock reduces freezing problems and grease contamination
- Coating is autoclavable to 121 °C (250 °F) moist heat or 110 °C (230 °F) dry heat
- Resistant to thermal shock
PYREXPLUS® labware allows for a better hand grip and clamp hold. PVC coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Can be written on with any standard lab marker, permanent or water-soluble. Do not use plastic-coated ware over direct or open flame.
4 Options Available Below
Product Details & Documents
PYREXPLUS®, borosilicate glass.
- Fluoropolymer resin stopcock reduces freezing problems and grease contamination
- Coating is autoclavable to 121 °C (250 °F) moist heat or 110 °C (230 °F) dry heat
- Resistant to thermal shock
PYREXPLUS® labware allows for a better hand grip and clamp hold. PVC coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Can be written on with any standard lab marker, permanent or water-soluble. Do not use plastic-coated ware over direct or open flame.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 89091-670
- 89091-696
- 30356-249
- 30356-252
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
- CapacityStopcock sizeStopper no.AvailabilityPrice
- Supplier Number: 66402-125Specifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89091-670Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 66402-500Specifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89091-696Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 66402-1LSpecifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.30356-249Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 66402-2LSpecifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.30356-252Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey