ContactUsLinkComponent
Erythroid Progenitor Reprogramming Kit
Erythroid Progenitor Reprogramming Kit
Catalog # MSPP-05924
CAS Number:  
undefined
Erythroid Progenitor Reprogramming Kit
Catalog # MSPP-05924
Supplier Number:  05924
CAS Number:  
Specifications
  • Description:
    Erythroid progenitor reprogramming kit
  • Size:
    1 kit
  • Cat. no.:
    MSPP-05924
Specifications
About this item
Integrated workflow containing tools to isolate and expand erythroid progenitor cells from peripheral blood and their subsequent reprogramming to iPS cells.
The erythroid progenitor reprogramming kit contains an integrated set of tools and reagents to enrich, expand, and reprogram erythroid progenitor cells from peripheral blood.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').