BrainPhys™ Imaging Optimised Medium
Catalog # MSPP-05796
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Description:BrainPhys™ Imaging optimized medium
- Size:500 ml
- Cat. no.:MSPP-05796
Specifications
About this item
Serum-free neurophysiological basal medium for improved neuronal live imaging and function.
BrainPhys™ Imaging optimized Medium is a serum-free neuronal basal medium that is optimized for imaging applications. Applications of BrainPhys™ Imaging optimized Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture. In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').