LabKlenz™ 200 Acid Detergents
Catalog # CA89089-212
Supplier: Steris
Specifications
- Size:3.8 L (1 gal.)
- Cat. no.:CA89089-212
- Packaging:Plastic bottle, 4/case
- Supplier No.:1L2008
Specifications
About this item
LabKlenz™ 200 Detergent is a low-foaming, phosphoric acid based liquid detergent.
- Flexibility - Effective removal of laboratory residues in both soft and hard water
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - Formulated with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 200 Detergent is specially formulated for use in automated glassware washing applications both as a rinse aid and for the removal of particulate, scale, and inorganic soils.