ContactUsLinkComponent
STEMdiff™ Microglia Maturation Kit
STEMdiff™ Microglia Maturation Kit
Catalog # MSPP-100-0020
CAS Number:  
undefined
STEMdiff™ Microglia Maturation Kit
Catalog # MSPP-100-0020
Supplier Number:  100-0020
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    STEMdiff™ microglia maturation kit
  • Size:
    1 kit
  • Cat. no.:
    MSPP-100-0020
Specifications
About this item
Maturation kit for the generation of microglia from human ES and iPS cell-derived microglia precursors.
The STEMdiff™ microglia culture system comprises STEMdiff™ microglia differentiation kit and STEMdiff™ microglia maturation kit. Together, these kits are used to differentiate and mature microglia derived from human pluripotent stem cells (hPSCs) using STEMdiff™ hematopoietic kit. Based on the protocol from the laboratory, the resulting cells are a highly pure population of microglia (>80% CD45/CD11b-positive, >50% TREM2-positive microglia; >20% morphologically distinct monocytes or macrophages). Cells derived using these products are versatile tools for modeling neuroinflammation, studying human neurological development and disease, co-culture applications, and toxicity testing.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').