ContactUsLinkComponent
MethoCult™ H4431 Medium
MethoCult™ H4431 Medium
Catalog # MSPP-04431
CAS Number:  
undefined
MethoCult™ H4431 Medium
Catalog # MSPP-04431
Supplier Number:  04431
CAS Number:  
Specifications
  • Description:
    MethoCult™ H4431
  • Size:
    100 ml
  • Cat. no.:
    MSPP-04431
Specifications
About this item
Methylcellulose-based medium with agar-LCM and EPO for human cells.
MethoCult™ H4431 is a methylcelluose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4431 supports the growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM). This formulation contains agar leukocyte conditioned medium (agar-LCM) as a source of colony-stimulating factors, plus recombinant human erythropoietin (EPO). It is suitable for use as a control medium for detection of 'EPO-independent' erythroid progenitor cells using MethoCult™ H4531.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').