Orders LinkContactUsLinkComponent
ASTM Class 7 Hooked Weights
Supplier:   Troemner
ASTM Class 7 Hooked Weights
Supplier:  Troemner
Certificates
ASTM Class 7 Hooked Weights
ASTM Class 7 Hooked Weights
7 Options Available Below
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
  • 303 stainless steel
  • ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Product Family Options
Product Information
  • Weight
    Availability
    Price
  • 0.1 N
    Log in for availability
    Log in for availability
    Each
    Each - $53.21
    $53.21
    New Customer Promotional Price
    Each
       $53.21
    Supplier Number: 30390784
  • 0.2 N
    Log in for availability
    Log in for availability
    Each
    Each - $53.21
    $53.21
    New Customer Promotional Price
    Each
       $53.21
    Supplier Number: 30390783
  • 0.5 N
    Log in for availability
    Log in for availability
    Each
    Each - $59.53
    $59.53
    New Customer Promotional Price
    Each
       $59.53
    Supplier Number: 30390782
  • 1 N
    Log in for availability
    Log in for availability
    Each
    Each - $66.71
    $66.71
    New Customer Promotional Price
    Each
       $66.71
    Supplier Number: 30390781
  • 2 N
    Log in for availability
    Log in for availability
    Each
    Each - $85.78
    $85.78
    New Customer Promotional Price
    Each
       $85.78
    Supplier Number: 30390780
  • 5 N
    Log in for availability
    Log in for availability
    Each
    Each - $132.52
    $132.52
    New Customer Promotional Price
    Each
       $132.52
    Supplier Number: 30390779
  • 10 N
    Log in for availability
    Log in for availability
    Each
    Each - $275.18
    $275.18
    New Customer Promotional Price
    Each
       $275.18
    Supplier Number: 30390778