To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Total hardness colormetric reagents
- Environmentally Preferable:
- Cat. no.:CA470350-700
- Range:0 - 250 mg/L, 200 - 500 mg/L, or 400 - 750 mg/L as CaCO₃ total hardness
- No. of Tests:100
Specifications
About this item
Reagents for low, medium and high range colorimetric determination of total hardness.
- The results will be displayed in mg/L (ppm) as CaCO₃
- These reagents are designed to be used with samples that have an expected range of 0 to 250 mg/L, 200 to 500 mg/L, or 400 to 750 mg/L
Depending on the range of the sample, one of three bottles of reagent A will be used, while reagent B and C are used for all ranges. By simply adding the range-appropriate bottle of liquid reagent A, two drops of reagent B, and the packet of reagent C to the sample, the reaction between calcium and magnesium and the reagents causes a color change dependent upon concentration. The intensity of color is determined by the compatible photometer and the concentration will be displayed.