ContactUsLinkComponent
Hanna Instruments NPK Soil Chemical Test Kit
  CA470349-848
 :  Hanna
New Product
 :  
undefined
Hanna Instruments NPK Soil Chemical Test Kit
  CA470349-848
 :  Hanna
 :  HI3895
New Product
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    NPK soil chemical test kit
  • Environmentally Preferable:
  • Cat. no.:
    CA470349-848
  • No. of Tests:
    10
This chemical test kit will determine the commonly monitored parameters in soil testing.
  • Pre-made reagents for ease of use
  • Measure pH for the most important elements of plant growth: Nitrogen, phosphorus and potassium
  • All reagents marked with expiration date and lot number for traceability
  • Test tubes for convenient handling
This test kit is supplied with all of the necessary reagents and equipment to perform each analysis, and all reagents are individually available as they run out. It provides users with a fundamental tool to assess important characteristics related to soil quality. All required materials are included with the test kit, such as the test tubes, color comparison cards, reagents and solutions. Methods include colorimetric for pH and nitrogen and turbidimetric for potassium.
 : This kit is equipped with all the necessary reagents and equipment to perform over 10 tests for each parameter. Replacement reagents sold separately.