Top Loading Vertical Lab Autoclaves, LabSci
Supplier: TUTTNAUER USA CO. LTD.
New Product
Certificates
About this item
Autoclave everything in your laboratory, including liquids. The LabSci Vertical lab autoclaves were developed specifically for educational institutions and research facilities. Designed with simplicity and economy in mind.
- High capacity
- Optimization of resources - water, power, operating time
- Self contained, no plumbing/drainage/venting needed
- Built in swivel wheels for portability
- Affordable and reliable solution for managing laboratory workload
- Sterilize liquids, glassware, unwrapped solids
Certifications: ISO 9001; ISO 14001; 2014/68/EU; ASME Section VIII Division 1; IEC 61010-1 / UL 61010-1; IEC 61010-2-040; EN 61326-1; ETSI EN 301 489-1; ETSI EN 301 489-17 certified.
Delivery: LabSci 60LV is supplied with two mesh wire baskets (77574-040) and LabSci 85LV is supplied with two mesh wire baskets (77574-038).
Specifications
- Catalog No:
- 77574-044
- 77574-042
- Max. working pressure:
- 40.6 psi
- 40.6 psi
Specifications
Related Information
Thermal printer
Product Information
Frequently Bought Together