Human Recombinant NRAS, Active (Biotinylated, His tag) (from E. coli), Biotinylated
New Product
Recombinant human NRAS (1-186) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
- Shipping temperature: Dry ice
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:NRAS1|N-ras|NS6|CMNS|NCMS|ALPS4
- Protein/peptide name:NRAS
- Molecular weight:~27 kDa
- Formulation:Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol.
- Tested applications:Enzyme assay
Frequently Bought Together