Human Recombinant HRAS (Mature form, His tag), Active (from E. coli)
New Product
Recombinant human HRAS (2-186) was expressed in E.coli cells using an N-terminal His tag.
- Shipping temperature: Dry ice
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:C-H-RAS|CTLO|p21ras|HRAS1|RASH1|C-HA-RAS1|H-RASIDX|HAMSV|C-BAS/HAS
- Protein/peptide name:HRAS
- Molecular weight:~23 kDa
- Formulation:Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol.
- Tested applications:WB, enzyme assay
Frequently Bought Together