Human Recombinant Rb (N-Term, GST tag) (from E. coli)
Supplier: Sino Biological
Certificates
About this item
Recombinant human Rb (46-378) was expressed in E. coli cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:pRb|OSRC|pp110|PPP1R130|RB|p105-Rb
- Protein/peptide name:RB1
- Molecular weight:~62 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Specifications
Frequently Bought Together