Order Entry
Canada
ContactUsLinkComponent
Human Recombinant PTPN11 (T253M Q257L, GST tag), Active (from Baculovirus-Insect Cells)
Human Recombinant PTPN11 (T253M Q257L, GST tag), Active (from Baculovirus-Insect Cells)
Supplier:  Sino Biological
New Product
Certificates
Human Recombinant PTPN11 (T253M Q257L, GST tag), Active (from Baculovirus-Insect Cells)
Supplier:  Sino Biological
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein synonyms:
    SHP2|PTP2C|PTP-1D|SH-PTP3|NS1|CFC|SH-PTP2|BPTP3
  • Protein/peptide name:
    PTPN11
  • Molecular weight:
    ~98 kDa
  • Formulation:
    Supplied as sterile 20 mM MOPS, pH 7.5, 300 mM NaCl, 10 mM glutathione, 0.25 - 1 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
  • Tested applications:
    WB, phosphatase assay

Specifications

Frequently Bought Together