Human Recombinant PTPN6 (SHP1, His tag), Active (from E. coli)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant full length human PTPN6 was expressed in E.coli cells using a C-terminal His tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:SH-PTP1|HPTP1C|HCPH|SHP-1L|SHP1|SHP-1|PTP-1C|HCP
- Protein/peptide name:PTPN6
- Molecular weight:67 kDa
- Formulation:Supplied as sterile 20 mM MOPS, pH 7.0, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.25 - 1 mM DTT, 10 - 25% glycerol.
- Tested applications:WB, phosphatase assay
Specifications
Frequently Bought Together