Human Recombinant PI3K (p110 alpha(H1047R, His tag)/p85 alpha, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant full length human p110 alpha (H1047R) and p85 alphasubunits were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein/peptide name:PIK3CA/PIK3R1
- Molecular weight:~124 kDa (p110), ~84 kDa (p85)
- Formulation:Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol.
- Tested applications:Kinase assay
Specifications
Frequently Bought Together