Human Recombinant FLT3 (Y591-V592insVDFREYEYD, GST tag), Active (from Baculovirus (Insect cells))
New Product
Recombinant human FLT3 (Y591-V592insVDFREYEYD) or FLT3-ITD ((571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:FLK-2|CD135|STK1|FLK2
- Protein/peptide name:FLT3
- Molecular weight:~76 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
- Tested applications:Kinase Assay
Frequently Bought Together