Human Recombinant HER2 (T798I, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant human HER2 (T798I) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:NEU|MLN19|HERVW|TKR1|HERV7Q|MLN 19|CD340|ERVWE1|HER2|HERV-W-ENV|ENV|HER-2/neu|HER-2|ENVW|HERVWENV|HERV-7q|NGL
- Protein/peptide name:ERBB2
- Molecular weight:~115 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
- Tested applications:Kinase Assay
Specifications
Related Information
Frequently Bought Together