Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant EGFR (L792H C797S L858R, GST tag), Active (from Baculovirus (Insect cells))
New Product
 
Human Recombinant EGFR (L792H C797S L858R, GST tag), Active (from Baculovirus (Insect cells))
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein synonyms:
    PIG61|HER1|mENA|ERBB|NISBD2|ERBB1
  • Protein/peptide name:
    EGFR
  • Molecular weight:
    89 - 100 kDa
  • Formulation:
    Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
  • Tested applications:
    Kinase Assay

 

Frequently Bought Together