One-Minute Competent Cells (DH5 Alpha)
About this item
The one-minute competent cells (DH5 Alpha) are engineered for chemical transformation with high efficiency.
- One-Minute competent cells are highly efficient (>10E8 transformants/µg pUC19)
- No need for heat shocking or followed shaking as traditional transformation
- One-minute processing time and directly spread on solid medium (agar plate)
- Broad utility for general cloning, blue-white screening, or plasmid isolation
- Multiple formats for high-throughput applications
Just mix DNA with this competent cell for 1 minute and spread on plate. No heat shock, no shaking needed. This product can be used for cloning, sub-cloning, synthesized DNA, or PCR fragment assembly, library construction and more. 50 µl per PCR tube.
Packaging: Shipped with dry ice for overnight delivery. Stored at –80 °C.
Caution: This product is intended for research use only.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The one-minute competent cells (DH5 Alpha) are engineered for chemical transformation with high efficiency.
- One-Minute competent cells are highly efficient (>10E8 transformants/µg pUC19)
- No need for heat shocking or followed shaking as traditional transformation
- One-minute processing time and directly spread on solid medium (agar plate)
- Broad utility for general cloning, blue-white screening, or plasmid isolation
- Multiple formats for high-throughput applications
Just mix DNA with this competent cell for 1 minute and spread on plate. No heat shock, no shaking needed. This product can be used for cloning, sub-cloning, synthesized DNA, or PCR fragment assembly, library construction and more. 50 µl per PCR tube.
Packaging: Shipped with dry ice for overnight delivery. Stored at –80 °C.
Caution: This product is intended for research use only.
Product Family Options
Product Information
- VolumeAvailabilityPrice
- 77574-024Competent cells, DH5 one-minuteSupplier Number: L2A1-8Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 77574-026Competent cells, DH5 one-minuteSupplier Number: L2A1-96Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)