ContactUsLinkComponent
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Supplier:  Cytiva
Certificates
undefined
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Supplier:  Cytiva
Specifications
  • Catalog No:
  • CA77548-832
  • CA77548-830
  • CA77548-836
  • CA77548-834
  • Type:
  • Syringe filter
  • Syringe filter
  • Syringe filter
  • Syringe filter
  • Hold-up volume:
  • <125 µl
  • <125 µl
  • <125 µl
  • <125 µl
  • Max. operating pressure:
  • 2.1 bar
  • 2.1 bar
  • 2.1 bar
  • 2.1 bar
  • Max. temperature:
  • 82 ℃
  • 82 ℃
  • 82 ℃
  • 82 ℃
  • Water flow rate:
  • 795 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 795 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 795 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 795 ml/min at 1.0 bar (100 kPa, 15 psi)