ContactUsLinkComponent
Terrific Agar
Terrific Agar
Catalog # 104028-320
Supplier:  Alkali Scientific
CAS Number:  
undefined
Terrific Agar
Catalog # 104028-320
Supplier:  Alkali Scientific
Supplier Number:  DC0135
CAS Number:  
Specifications
  • Description:
    CellPro™ terrific agar
  • Cat. no.:
    104028-320
  • Plate Diameter:
    100 mm
Specifications
About this item
Support bacterial growth with Terrific Agar, providing essential nutrients for robust colony formation and culture.
  • 1.20% Tryptone
  • 2.40% Yeast Extract
  • 1.60% Glycerol
  • 1.25% Potassium Phosphate Dibasic
  • 0.23% Potassium Phosphate Monobasic
  • 1.50% Agar
Terrific Agar is a specialized culture medium used for the propagation of recombinant bacterial strains, particularly Escherichia coli (E. coli), in molecular biology laboratories. This agar medium is a modified version of Terrific Broth (TB) agar, supplemented with glycerol, yeast extract, tryptone, and phosphate buffer. Terrific Agar provides optimal conditions for high-density bacterial growth and protein expression, making it ideal for protein production and purification studies. The plates (Ø 100 mm) are convenient for routine bacterial culture experiments and can support the growth of transformed bacterial cells carrying recombinant DNA constructs.