CellPro™ LB (Lennox) + Chloramphenicol
Supplier: Alkali Scientific
Certificates
About this item
Maintain optimal bacterial growth with CellPro™ LB (Lennox) + Chloramphenicol, ensuring reliable results in your molecular biology experiments.
- Enhance selection with a potent combination of Lennox LB broth and chloramphenicol
- Ensure efficient bacterial growth and selection for your experiments
- Rely on consistent performance for reliable research outcomes
CellPro™ LB (Lennox) + Chloramphenicol is a specialized culture medium formulated for the growth of bacterial strains resistant to chloramphenicol. This LB agar variant contains chloramphenicol, a bacteriostatic antibiotic that inhibits protein synthesis in susceptible bacteria. By incorporating chloramphenicol into the LB agar, this medium selectively promotes the growth of chloramphenicol-resistant bacterial strains, making it ideal for the maintenance of recombinant DNA constructs and other molecular biology applications.
Frequently Bought Together