About this item
Conduct sterile cell culture experiments with Erlenmeyer Flasks, PC Conical with Baffles, provided sterile for your convenience.
- Sterilized by E-Beam
- Non pyrogenic
- Individually wrapped in a plastic bag
- Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination
These CellPro™ Erlenmeyer Flasks, crafted from durable polycarbonate (PC) material, feature a conical design with strategically placed baffles, ensuring efficient mixing and aeration essential for successful cell culture applications. This sterile flask, meticulously manufactured and individually packaged, offers researchers a reliable and contamination-free vessel for their cell culture experiments. With its robust construction, the flask provides stability and longevity, while its sterile nature ensures consistency and reproducibility in research outcomes. Whether used for routine cell culture or specialized applications, the CellPro™ Erlenmeyer Flask stands as a trusted companion in the laboratory, facilitating precise and reliable results in molecular biology studies.