ContactUsLinkComponent
CellPro™ MOPS Buffer 10X, Molecular Biology Grade, Sterile Filtered
CellPro™ MOPS Buffer 10X, Molecular Biology Grade, Sterile Filtered
  104028-064
 :  
undefined
CellPro™ MOPS Buffer 10X, Molecular Biology Grade, Sterile Filtered
  104028-064
 :  DB0356-CS
 :  
  • Description:
    CellPro™ MOPS, 10X, brown plastic bottle
  • Size:
    1 L, 12/case
  • Cat. no.:
    104028-064
Prepare buffers for electrophoresis with CellPro™ MOPS Buffer 10X, meeting molecular biology grade standards and sterile filtration.
  • Optimized for RNA agarose electrophoresis
  • Ideal for use with RNA formaldehyde gels
CellPro™ MOPS Buffer 10X is a versatile reagent commonly used in molecular biology and biochemistry applications. With its optimized formulation, this buffer provides stable pH conditions, crucial for nucleic acid electrophoresis, RNA isolation, and protein gel electrophoresis. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ MOPS Buffer 10X a trusted choice for researchers seeking reliable results in their experiments.