Order Entry
Canada
ContactUsLinkComponent
 
CellPro™ TBS, 20X, pH 7.4, Sterile
Catalog # 104027-468
Supplier:  Alkali Scientific
CAS Number:  
undefined
CellPro™ TBS, 20X, pH 7.4, Sterile
Catalog # 104027-468
Supplier:  Alkali Scientific
Supplier Number:  DB0246
CAS Number:  

Specifications

  • Concentration:
    20X
  • Description:
    CellPro™ TBS
  • Size:
    2000 ml
  • Cat. no.:
    104027-468
  • pH:
    7.4

Specifications

About this item

Prepare reliable Tris-buffered saline with CellPro™ TBS, available in 20X concentration, pH 7.4, sterile and filtered.

  • Space-saving stock solution
  • Ideal for preparing TBS or TBST wash buffers or blocking buffer diluent
  • For ELISA, Western blotting and other immunoassay applications

CellPro™ TBS, 20X, pH 7.4, offers a sterile and concentrated Tris-buffered saline (TBS) solution, meticulously formulated to meet the demands of various molecular biology applications. With a pH of 7.4, this buffer provides optimal conditions for a wide range of biochemical and immunological assays, ensuring reliable and reproducible results. The 20X concentration allows convenient dilution to the desired working strength, providing flexibility and efficiency in the laboratory. Sterilized to maintain purity and eliminate the risk of contamination, CellPro™ TBS is ready for immediate use in your research experiments. Trust in the quality and consistency of CellPro™ buffers to support your molecular biology endeavors, delivering accurate and precise results every time.