Order Entry
Canada
ContactUsLinkComponent
CellPro™ BSA-Blocking Buffer, 3% + Tween-20 0.05% in TBS
CellPro™ BSA-Blocking Buffer, 3% + Tween-20 0.05% in TBS
Catalog # 104028-342
Supplier:  Alkali Scientific
CAS Number:  
undefined
CellPro™ BSA-Blocking Buffer, 3% + Tween-20 0.05% in TBS
Catalog # 104028-342
Supplier:  Alkali Scientific
Supplier Number:  DB0228
CAS Number:  

Specifications

  • Description:
    CellPro™ BSA-blocking buffer, 3% + Tween-20 0.05% in TBS
  • Size:
    250 ml
  • Cat. no.:
    104028-342

Specifications

About this item

Enhance protein detection with CellPro™ BSA-Blocking Buffer, featuring 3% BSA and 0.05% Tween-20 in TBS for effective blocking.

  • BSA (bovine serum albumin) blocking buffer is used as a blocking reagent ideal for saturating excess protein-binding sites on membranes and microplates for Western blotting and ELISA applications

CellPro™ BSA-Blocking Buffer presents a meticulously formulated solution essential for immunoassays and protein-related research endeavors. This 3% bovine serum albumin (BSA) solution, augmented with 0.05% Tween-20 in Tris-buffered saline (TBS), serves as a pivotal blocking agent to prevent non-specific binding in various biochemical applications. With a standardized concentration and meticulously balanced composition, this buffer ensures consistent and reliable results, minimizing background noise and enhancing assay sensitivity.

Its compatibility with a wide range of immunoassays, including Western blotting, ELISA, and immunohistochemistry, underscores its versatility and utility in diverse laboratory settings. Packaged in a convenient 250 ml volume, this CellPro™ BSA-Blocking Buffer streamlines experimental workflows, providing researchers with a reliable tool to optimize assay performance and achieve accurate results. Trust in the quality and consistency of CellPro™ BSA-Blocking Buffer to support your research endeavors and drive scientific discovery forward.