Specifications
- Concentration:0.2 M
- Description:CellPro™ Phosphate buffer
- Size:1 L
- Cat. no.:104026-972
- pH:7.0
Specifications
About this item
Ensure reliable results in molecular biology applications with sterile and filtered CellPro™ Phosphate Buffer.
- High-purity solutions available in a wide range of grades
- Multiple and user-friendly packaging options
- Assured solution stability with pH control
- No interference with biological processes
- Scalable from early research to commercial applications
- Customized blending
CellPro™ Phosphate Buffer offers a sterile and filtered solution vital for numerous laboratory applications. With its meticulously prepared formulation, this buffer ensures purity and consistency, meeting the stringent requirements of scientific research and experimentation. The sterile nature of the solution minimizes the risk of contamination, making it ideal for various biochemical and molecular biology techniques. Whether utilized for protein assays, enzyme activity studies, or cell culture maintenance, CellPro™ Phosphate Buffer provides a reliable and standardized solution for diverse laboratory needs. Its filtered formulation guarantees clarity and reliability in experimental results, providing researchers with confidence in their findings. With CellPro™ Phosphate Buffer, scientists can achieve consistent performance and reproducible results, supporting the success of their research endeavors.