ContactUsLinkComponent
Liquid Nitrogen Storage System
  104027-600
undefined
Liquid Nitrogen Storage System
  104027-600
 :  LN5
  • Capacity:
    60 L
  • Description:
    Liquid nitrogen storage system
  • Cat. no.:
    104027-600
Store biological samples safely with the liquid nitrogen storage system, providing reliable cryogenic storage solutions.
  • Easy-access lids
  • Convenient inventory management options
  • Streamline sample retrieval and organization
  • 60 L capacity
The liquid nitrogen storage system is a robust and reliable solution designed to safely store biological samples at ultra-low temperatures. With its advanced design and construction, this system ensures the long-term preservation of valuable specimens, such as cells, tissues, and biological fluids. Equipped with durable insulation materials and a secure locking mechanism, it effectively maintains the temperature stability required for cryogenic storage, even in demanding laboratory environments. It is used for research, clinical, or industrial applications, the liquid nitrogen storage system provides researchers and technicians with peace of mind, knowing that samples are stored securely and maintained at optimal conditions for future analysis and experimentation.