CellPro™ LB (Lennox), Molecular Biology Grade, Sterile Filtered
Ensure consistent bacterial culture results with CellPro™ LB (Lennox), provided sterile filtered and meeting molecular biology grade standards.
- For molecular biology
- For maintaining and propagating E.coli culture
CellPro™ LB (Lennox) is a molecular biology-grade growth medium designed for the cultivation of Escherichia coli strains. With its balanced composition of tryptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to maintain purity and consistency, ensuring optimal performance in bacterial culture, plasmid DNA isolation, and recombinant protein production.
Frequently Bought Together