CellPro™ Citrate Buffer, Sterile
104028-998
:
- Pack type:1 L, 12/case
- Description:CellPro™ Citrate buffer, 10 mm
- Cat. no.:104028-998
- pH:6.0
Prepare for molecular biology assays with CellPro™ citrate buffer, provided sterile for your convenience.
- Sterile
Introducing CellPro™ Citrate buffer, meticulously prepared to meet the stringent requirements of your laboratory experiments. This sterile buffer solution is crafted with precision to ensure optimal performance and reliability in a variety of applications. Whether used in molecular biology, biochemistry, or cell culture, CellPro™ citrate buffer offers consistent quality and purity, providing the ideal environment for your research needs.