CellPro™ 15 ml Conical Centrifuge Tubes, Polypropylene, Conical Bottom
Catalog # 104027-466
Supplier: Alkali Scientific
Specifications
- Sterility:—
- Description:15 ml centrifuge tube rack, fits 50×15 ml
- Cat. no.:104027-466
- Packaging:1 per pack
Specifications
About this item
Facilitate efficient sample processing with CellPro™ 15 ml conical tubes, made of polypropylene with a conical bottom for optimal centrifugation.
- Sterile
- RNase, DNase and pyrogen free
- Polypropylene (PP)
- Marker caps
- Oversized white ID patch on sidewall
CellPro™ 15 ml Marker centrifuge tubes feature a unique white Marker screw cap that is easy to write on with a Sharpie or similar lab marker and allows easy identification of sample on the top. They also have easy-to-read graduations and an oversized white ID patch on the sidewall. The tubes are RNase, DNase and pyrogen free, and come packaged either in green lab certified recyclable sterile racks or resealable sterile bags. CellPro™ marker tubes are highly resistant to organic solvents and exhibit a stable sample collection and processing environment for nearly all liquids and solids.