ContactUsLinkComponent
50 ml Centrifuge Tubes, Conical, Bagged, Sterile, PP, Free Standing (500 Per Case)
50 ml Centrifuge Tubes, Conical, Bagged, Sterile, PP, Free Standing (500 Per Case)
  104027-974
undefined
50 ml Centrifuge Tubes, Conical, Bagged, Sterile, PP, Free Standing (500 Per Case)
  104027-974
 :  C5615
  • Sterility:
    Sterile
  • Description:
    15 ml Centrifuge tubes, PP (17×118 mm), flat screw cap, individually wrapped
  • Cat. no.:
    104027-974
  • Packaging:
    Individually wrapped
Streamline your sample preparation with sterile 50 ml conical centrifuge tubes, bagged and free-standing in a case of 500.
  • Sterile, RNase, DNase, pyrogen and cytotoxin free
  • Available racked or bagged
  • Compatible with standard rotors
  • Frosted marking surface
Experience optimal sample handling with CellPro™ 50 ml centrifuge tubes. Designed with convenience and efficiency in mind, these conical tubes are bagged, sterile and crafted from premium polypropylene. Their free-standing capability ensures stability during centrifugation. Each case contains 500 tubes, offering a sizable supply to meet the demands of your laboratory operations.