CellPro™ LB Lennox Agar
Supplier: Alkali Scientific
Certificates
About this item
Promote bacterial growth with CellPro™ LB lennox agar, a high-quality medium suitable for various microbiology applications.
- Enhance bacterial culturing with CellPro™'s specialized LB lennox agar
- Ensure robust growth and colony formation for precise microbiology studies
- Trust in consistent performance and reliable results for your research endeavors
CellPro™ LB lennox agar is a modification of Luria-Bertani (LB) agar, formulated with reduced sodium chloride content compared to standard LB agar. This medium is commonly used for molecular biology applications, including the cultivation and selection of recombinant bacterial strains. LB lennox agar provides a nutrient-rich environment for bacterial growth, supporting the propagation of transformed cells harboring plasmids with antibiotic resistance markers.
Frequently Bought Together