Specifications
- No. of wells:384
- Description:Cell culture plates with lids, non-treated
- Cat. no.:104027-190
- Packaging:50/case
- Sterile:Sterile
Specifications
About this item
Cultivate cells with ease using CellPro™ flat bottom, non-treated cell culture plates, equipped with lids for secure containment of samples.
- High clarity, 100% virgin polystyrene
- Vacuum gas plasma tissue culture treatment
- Clear lot number for batch tractability
- Peel-to-open, individually wrapped and sterilized by gamma irradiation
- Markings of well coordinates available for 12, 24, 48 and 96 well plates
- Non-pyrogenic, DNase/RNase free
- Gamma sterilized
CellPro™ flat bottom cell cultureplates, designed for a wide range of cell culture applications. These plates feature a flat bottom surface, providing optimal stability for cell growth and attachment. Each plate comes with a matching lid to prevent contamination and evaporation, ensuring the integrity of your cell cultures. Manufactured from high-quality materials, these non-treated plates offer versatility for use with various cell lines and culture media.
CellPro™ flat bottom cell culture plates are used for performing routine cell culture experiments or specialized assays with consistent and reproducible results.