ContactUsLinkComponent
Immunoplates, 96 well, ELISA Maxibinding, Polystrene, Flat Bottom with Lid
Immunoplates, 96 well, ELISA Maxibinding, Polystrene, Flat Bottom with Lid
  104028-404
undefined
Immunoplates, 96 well, ELISA Maxibinding, Polystrene, Flat Bottom with Lid
  104028-404
 :  MPMAXI
  • Color:
    Clear
  • Description:
    96 well microplate, maxibinding, F-type
  • Material:
    PS
  • Size:
    85.4×127.6 mm
  • Cat. no.:
    104028-404
  • Packaging:
    10/sleeve, 100/case
Perform ELISA assays efficiently with Immunoplates, featuring a clear PS construction, 96 wells, flat bottom with lid, available in cases of 100.
  • Clear polystyrene
  • Surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assays
  • Alphanumerically labeled rows and columns
Maximize the efficiency of ELISA assays with the Immunoplates. Constructed from clear polystyrene, these 96-well plates feature a flat bottom design and come with lids for convenient sample protection. The ELISA MAXIBINDING surface ensures optimal antibody binding and assay sensitivity. Each case contains 100 plates, providing a dependable solution for high-throughput ELISA applications. Trust in the superior quality and performance of these immunoplates to streamline your laboratory workflows and achieve accurate results.