Hand Sanitizer Dermasan 70% Alcohol Gel, with Pump
104026-846
- Capacity:1 ml
- Description:Hand sanitizer gel with pump, 1 gal.
- Cat. no.:104026-846
Maintain hand hygiene standards with Hand Sanitizer Dermasan 70% alcohol gel, packaged in a convenient pump dispenser according to W.H.O. guidelines.
- Pump included
- 70% alcohol gel
Dermasan Hand Sanitizer is a premium-grade alcohol-based gel formulated to effectively sanitize hands and surfaces, meeting the guidelines set by the World Health Organization (W.H.O.). With a potent 70% alcohol content, this sanitizer provides powerful germ-killing action against bacteria, viruses, and other pathogens. The convenient pump dispenser included with the 1 gallon packaging ensures easy and mess-free application, making it suitable for use in healthcare facilities, offices, schools, and households.