Specifications
- No. of wells:384
- Sterility:Sterile
- Color:Clear
- Description:Cell culture plate, TC
- Cat. no.:104028-602
- Packaging:1 per pack, 100 per case
- Bottom Style:Flat bottom
Specifications
About this item
Achieve high-throughput cell culture with the Alkali Scientific™ 384 well cell culture plates, featuring a flat bottom design.
- Total well volume of 112 μl; working well volume of 20 to 80 μl
- Cell culture microplates are sterile and non pyrogenic
- The 384-well universal optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties
Introducing Alkali Scientific's™ 384-well cell culture plates, meticulously designed to meet the demands of high-throughput cell culture applications with unparalleled efficiency and reliability. Featuring a flat bottom design, these plates provide optimal cell attachment and growth, ensuring uniform and consistent results across all wells. Engineered with premium-grade materials, these cell culture plates guarantee superior quality and performance, making them ideal for a wide range of cell-based assays, screenings, and experiments. Experience the utmost convenience and precision in your research endeavors with Alkali Scientific's™ 384-well cell culture plate.