Order Entry
Canada
ContactUsLinkComponent
Radiant™ SYBR Green Master Mix qPCR Kit, Lo-ROX
Radiant™ SYBR Green Master Mix qPCR Kit, Lo-ROX
Catalog # 104028-810
Supplier:  Alkali Scientific
CAS Number:  
undefined
Radiant™ SYBR Green Master Mix qPCR Kit, Lo-ROX
Catalog # 104028-810
Supplier:  Alkali Scientific
Supplier Number:  QS1020
CAS Number:  

Specifications

  • Description:
    Radiant™ Green Lo-ROX qPCR kit, (20×1 ml)
  • Cat. no.:
    104028-810
  • No. of Reactions:
    2000 reactions

Specifications

About this item

Utilize Alkali Scientific™ Radiant™ SYBR Green Master Mix qPCR Kit, Lo-ROX, for accurate quantitative PCR assays using SYBR Green detection with low ROX content.

  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Broad range detection for increased reliability in low-copy assays
  • Versatile 2X Mixes, suitable for both ultra-fast and standard cycling methods

Alkali Scientific offers Radiant™ Green Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Green Lo-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.

Instrument Compatibility:
The Radiant™ Green Lo-ROX qPCR Kit has been optimized for use optimized for use with intercalating-dye based real-time PCR on qPCR instruments that require a standard, low concentration of ROX such as Applied Biosystems® 7500, 7500 FAST, ViiA7, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, 6 Flex, 6 Pro QuantStudio™ 7, 7 Flex, 7 Pro, QuantStudio™ 12K Flex, QuantStudio™ DX and Stratagene (Agilent) ® MX4000, MX3000P, MX3005P. In addition, the Radiant™ Green Lo-ROX qPCR Kit is optimal for use on qPCR instruments that do not require the use of ROX for data normalization such as Bio-Rad Opticon™, Opticon2™, MiniOpticon, Chromo4™, CFX96, CFX384, Roche LightCycler®480, Qiagen Rotor-Gene™ 3000 and 6000, Eppendorf Mastercycler®ep realplex, Cepheid SmartCycler™, and Illumina® Eco™.