ProCycle™ Half-Skirted PCR Plates, Raised Rim, ABI®-Type, 96×0.2 ml Well
104028-554
- No. of wells:96
- Skirt:Semi-skirted
- Description:ProCycle™ mini skirted PCR plates, raised rim, 96×0.2 ml well, blck printed alphanumerics, A12 cut corner, clear, 10 per pack
- Cat. no.:104028-554
Optimize your PCR experiments with Alkali Scientific™ ProCycle™ half-skirted PCR plates, featuring raised rim design, ABI®-Type, 96×0.2 ml well, black printed alphanumerics, and A12 notch, available in packs of 10.
- Easy well identification: High-contrast, black alphanumeric inde×ing facilitates quick and accurate well identification, streamlining your workflow
- Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
- Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving sample integrity
- High clarity, ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
- Purity assurance: Plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free
Enhance your PCR workflow with ProCycle™ Half-Skirted PCR Plates. Featuring a raised rim design and ABI®-Type configuration, these plates provide optimal sample retention and compatibility with standard PCR instruments. The black printed Alphanumerics ensure easy well identification, streamlining your experimental setup. With a convenient A12 notch for orientation, these plates offer hassle-free handling and compatibility with automated systems.
: Each pack contains 10 plates, providing ample supply for your laboratory needs.